Imprimer
VWR® Adapters with Stopcock and Connector
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
201-1374EA 28.5 EUR
201-1374 201-1375 201-1376 201-1377 201-1378 201-1379 201-1396 201-1397
VWR® Adapters with Stopcock and Connector
Adapters Adapters, Joints with Tube Connectors
Borosilicate 3.3 glass.
  • Highly resistant to aggressive chemicals and thermal shock
  • Straight or right angled tubing connector
  • Standard ground joint and standard ground joint stopcock (NS)
  • Stopcock with solid glass key, bore 2,5 mm
Outer diameter of the tubing connector: 8 mm