Imprimer
VWR®, Dimroth Condensers with Standard Ground Joints
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
201-1409EA 67.7 EUR
201-1409 201-1412 201-1415
VWR®, Dimroth Condensers with Standard Ground Joints
Condensers
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • Standard ground cone and socket
These Dimroth condensers are equipped with a double-spiral for effective condensation of vapours. With GL 14 screw thread and 8 mm external diameter tubing connection.