Imprimer
Detection window maker for capillary electrophoresis
Fournisseur: MICROSOLV
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
07201-S 07204-S 07207-S
MICS07201-SEA 264 EUR
MICS07201-S MICS07204-S MICS07207-S
Detection window maker for capillary electrophoresis
Systèmes d'électrophorèse Capillaires pour électrophorèse
Creates a perfect detection window. Removes the polyimide coating from 365 µm OD capillaries for detection by UV and LIF.
  • Easy to use for all capillaries for CE
  • Takes 10 seconds to make a perfect window
The Window Maker™ is designed to produce enough heat to burn off the polyimide but not damage the glass in any way. Can be used to remove 1 to 2 mm of the polyimide from the inlet and outlet tips of the capillary.
Informations de livraison: Supplied with a standard 2 mm module for burning the capillary. Other size heating modules are available as accessories.