Imprimer
Cut resistant gloves, uvex unidur 6641
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6021010 6021006 6021007 6021009 6021008 6021011
UVEX6021010EA 154 EUR
UVEX6021010 UVEX6021006 UVEX6021007 UVEX6021009 UVEX6021008 UVEX6021011
Cut resistant gloves, uvex unidur 6641
Gants
These lightweight high-quality HPPE assembly gloves with PU coating on palm and fingertips provide level 3/B cut protection and a high level of sensitivity. It is perfectly suited to precision assembly work requiring tactility and cut protection.
  • Outstanding mechanical abrasion resistance
  • Good grip in dry and slightly damp areas
  • High flexibility
  • Very good tactile feel
  • Outstanding wearer comfort
Typical applications include: Automotive industry, maintenance, assembly, metal working and packaging.
Certifications: EN 388:2016 (4343B).