Imprimer
ESD safety ankle boots, lace-up, uvex 2, 6509
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6509.2/43 6509.2/39 6509.2/47 6509.2/35 6509.2/38 6509.2/46 6509.2/52 6509.2/44 6509.2/40 6509.2/37 6509.2/45 6509.2/41 6509.2/49 6509.2/50 6509.2/48 6509.2/51 6509.2/36 6509.2/42
UVEX6509.2/43EA 146 EUR
UVEX6509.2/43 UVEX6509.2/39 UVEX6509.2/47 UVEX6509.2/35 UVEX6509.2/38 UVEX6509.2/46 UVEX6509.2/52 UVEX6509.2/44 UVEX6509.2/40 UVEX6509.2/37 UVEX6509.2/45 UVEX6509.2/41 UVEX6509.2/49 UVEX6509.2/50 UVEX6509.2/48 UVEX6509.2/51 UVEX6509.2/36 UVEX6509.2/42
ESD safety ankle boots, lace-up, uvex 2, 6509
Chaussures et bottes
Metal-free, extremely lightweight, flexible, multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, and a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock-absorbent at heel and front of the foot
  • Virtually seamless design eliminates potential pressure points and blisters
  • Elastic laces for quick, individual adjustment
  • 100% metal-free xenova® toecap, non magnetic (200 J) anatomical shape, good lateral stability, no thermal conductivity
  • Flexible, metal-free, puncture resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black
Width: 11
Certifications: EN 61340 (ESD); EN ISO 20345: 2011, S3, SRC