Imprimer
M Stand for Bio-bin® Boxes
Fournisseur: ECONIX LTD
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BEXBBMS5.0 EXBBMS30.0
119-0468EA 429 EUR
119-0468 ECONEXBBMS30.0
M Stand for Bio-bin® Boxes
Boxes Waste Disposal Boxes
These stainless steel stands were designed to allow the 2, 5 and 30 Litre Bio-bin® boxes to tilt at approximately 15 degrees to allow easy filling for tall thin items e.g. pipettes.
  • Allow the user to see the contents easily while helping to prevent overfilling and also 'wigwamming' of items
  • Small stand for 2 and 5 L boxes is ideal for use on work benches
  • Larger 30 L M stand is perfect for floor use