Imprimer
ESD safety shoes, lace-up, uvex motion 3 XL S3, 6496
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6496.3/47 6496.3/48 6496.3/49 6496.3/50 6496.3/41 6496.3/42 6496.3/43 6496.3/44 6496.3/45 6496.3/46 6496.3/35 6496.3/36 6496.3/37 6496.3/38 6496.3/39 6496.3/40
UVEX6496.3/47EA 742 EUR
UVEX6496.3/47 UVEX6496.3/48 UVEX6496.3/49 UVEX6496.3/50 UVEX6496.3/41 UVEX6496.3/42 UVEX6496.3/43 UVEX6496.3/44 UVEX6496.3/45 UVEX6496.3/46 UVEX6496.3/35 UVEX6496.3/36 UVEX6496.3/37 UVEX6496.3/38 UVEX6496.3/39 UVEX6496.3/40
ESD safety shoes, lace-up, uvex motion 3 XL S3, 6496
Chaussures et bottes
Metal-free, extra high and wide safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar and a PUR outsole suitable for customised shoe adjustments
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Customised uvex EVA insole
  • Ample room for inserting special diabetic insoles
  • Individually adjustable elastic laces with quick-lock mechanism
  • Anti-twist heel cap for better stability and protection from ankle twisting
  • Virtually seamless design to eliminate the potential for pressure points and blisters
  • 100% metal-free uvex xenova® toe cap
  • Puncture resistant insole
  • Meets ESD requirement: resistance to ground < 35 megaohms
Suitable for people allergic to chrome, made from synthetic textiles.
Colour: Black
Width>15
Certifications: EN 61340 (ESD), EN ISO 20345:2011; S3, SRC