Imprimer
Chemical resistant gloves, uvex rubiflex S NB60S
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9893056 9893060 9893199
UVEX9893056EA 632 EUR
UVEX9893056 UVEX9893060 UVEX9893199
Chemical resistant gloves, uvex rubiflex S NB60S
Gants
These gauntlet style chemical protection gloves feature a reinforced cotton interlock lining and a full NBR coating. This makes the uvex Rubiflex S suitable for many applications in which protection against chemicals as well as increased mechanical protection are required.
  • Good resistance to chemicals, acids, alkalis, mineral oils and solvents
  • Outstanding wearer comfort and ergonomic fit
  • High flexibility
  • Silicone-free according to imprint test
Ideal for use in chemical industry, sewer construction, municipal cleaning and sandblasting.
Certifications: EN ISO 374-1:2016 Type B (JKOPT), EN 388:2016 (2121X)
Food contact approved