Imprimer
Cut resistant gloves, uvex unidur 6643
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6031410 6031408 6031409 6031407
UVEX6031410EA 149 EUR
UVEX6031410 UVEX6031408 UVEX6031409 UVEX6031407
Cut resistant gloves, uvex unidur 6643
Gants
These sensitive cut protection gloves with HPPE fibres and NBR coating on palm and fingertips are lightweight and provide a cut protection level of 3/B. They are also extremely sensitive.
  • Very high abrasion, tear and puncture resistance
  • Good grip in damp and oily areas
  • High flexibility and good tactile feel
  • Good wearer comfort
Ideal for use in the automotive and sheet metal industry, assembly, repair and maintenance work.
Certifications: EN 388:2016 (4344B)