Imprimer
Chemical resistant gloves, uvex rubiflex S NB35B
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6022407 6022408 6022409 6022410 6022411
UVEX6022407EA 152 EUR
UVEX6022407 UVEX6022408 UVEX6022409 UVEX6022410 UVEX6022411
Chemical resistant gloves, uvex rubiflex S NB35B
Gants
These cotton interlock chemical safety gloves feature a special NBR coating and combine protection with outstanding wearer comfort and flexibility.
  • Good mechanical properties
  • Good resistance to many chemicals
  • Very good grip in damp and wet areas
  • Outstanding tactile feel
  • Ergonomic fit, extremely high flexibility
  • Good wearer comfort with high water vapour absorption due to the cotton lining
Typical applications include: Automotive sector, chemical industry, laboratories, varnishing, food processing.
Certifications: EN 388:2016 (2111X), EN ISO 374-1:2016 Type A (JKNOPT)
OEKO-TEX® Standard 100