Imprimer
Cut resistant gloves, uvex synexo impact 1
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6059810 6059811
UVEX6059810EA 28.2 EUR
UVEX6059810 UVEX6059811
Cut resistant gloves, uvex synexo impact 1
Gants
These HPPE/glass fibre cut protection gloves with NBR coating and Grip finish on palm and fingertips combine the benefits of seamless safety gloves with specially designed protection zones.
  • Protection against shocks and impacts thanks to the extra padding in the palm area
  • Very high level of cut protection
  • Good grip in dry and damp areas
  • Good fit, high flexibility, good wearer comfort
  • Flexible velcro fastening
  • Protectors on the back of the hand and reinforcements on the finger joints
Ideal for use in tasks with extreme mechanical stress, handling heavy-duty tools, building and consctruction industry, mining, drilling, oil and gas industry.
Certifications: EN 388:2016 (4X43CP)