Imprimer
Homogeniser, Dounce Type
Fournisseur: BELLCO GLASS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1984-10007 1984-10100 1984-10040 1984-10002
BELC1984-10007EA 802 EUR
BELC1984-10007 BELC1984-10100 BELC1984-10040 BELC1984-10002
Homogeniser, Dounce Type
Homogénéisateurs Broyeurs pour tissus
Dounce tissue homogeniser designed for disruption of whole cells leaving cell nuclei undamage.
  • Polished tube and pestles
  • Made up of glass
Informations de livraison: Two pestles supplied with each unit, large clearance pestle A for initial processing and small clearance pestle B completes homogenisation.