Imprimer
Cut resistant gloves, uvex C500 M foam
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6049808 6049807 6049809 6049810 6049811
112-5368EA 240 EUR
112-5368 112-5367 112-5369 112-5370 112-5371
Cut resistant gloves, uvex C500 M foam
Gants
These bamboo-rayon/glass/polyamide/HPPE gloves feature a HPE SoftGrip foam coating on palm and fingers. The Bamboo TwinFlex® technology ensures maximum cut resistance, exceptional abrasion resistance and outstanding wearing comfort. Suitable for dry areas and slightly oily/damp areas.
  • Resist contact heat up to 100 °C
  • High flexibility
  • Highest cut resistance level
  • Silicone-free according to imprint test
  • Perfect fit with 3D Ergo man mould technology
  • Partially reinforced thumb joints
  • Crouch zone reinforcement, knitted cuff
Typical applications include: Automotive industry, construction, brewery/beverage industry, glass industry, repair work, metal working industry.
Certifications: EN 388:2016 (4X42C); EN 407 (X1XXXX)