Imprimer
Acide chlorhydrique dans l'éthanol pour la microscopie, Sigma-Aldrich®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): Hydrochloric acid solution, Muriatic acid
M-Clarity™ (Niveau de Qualité) = MQ300
1.00327.1000 1.00327.5000
1.00327.1000EA 62.2 EUR
1.00327.1000 1.00327.5000
Acide chlorhydrique dans l'éthanol pour la microscopie, Sigma-Aldrich®
Acide chlorhydrique
Hydrochloric acid in ethanol is used for the microscopical investigation of mycobacteria in sample material of human origin. It can be applied as decolorizing solution, as e.g. in the Tb-color staining set.
Once stained, the acid fast mycobacteria keep their coloring even after treatment with strong decolorizing solutions as HCl-ethanol. They remain stained red after counterstaining, whereas the microorganisms susceptible to acid take on the color of the counterstain. 1l of the Hydrochloric acid in ethanol is capable of decolorizing up to 500 slides.
The product is registered as IVD / CE product and can be used in diagnostics and laboratory accreditation.
Formule: HCl
Poids moléculaire: 36.46 g/mol
Densité: 0.90 g/cm3 (20 °C)
Point éclair: 21 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00011324
Numéro CAS: 7647-01-0
UN: 2924
ADR: 3 (8) II
IVDLogo