Imprimer
Microtitre tubes, 1,2 ml
Fournisseur: BIOZYM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
710410 710414
BZYM710410EA 0 EUR
BZYM710410 BZYM710414
Microtitre tubes, 1,2 ml
Tubes Microtubes
These standard 1,2 ml microtiter tubes are made of polypropylene and have a labeling field.
  • Autoclavable
  • Freezable
  • Can be closed with PlugStrip cap strips
The standard racks can be refilled easily and inexpensively with the reload system. Rack version works on Beckman, Tecan and Tomtec systems.
For individual vessels, the PlugStrip closures can be individually tailored. All microtiter tubes have an additional filling space that allows sample dilutions and/or sample mixing prior to transfer to microplates.
Possible applications include: HTLV-III tests, RIA, transport of specimen and reagents.
Informations de commande: Available in sachets as well as pre-fitted in 8×12 format. Also available is a robot-compatible rack version.