Imprimer
General purpose gloves, Showa 350R
Fournisseur: Showa
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
350R-10/XL 350R-7/S 350R-8/M 350R-9/L
112-5788EA 85.8 EUR
112-5788 112-5789 112-5790 112-5791
General purpose gloves, Showa 350R
Gants
These hand-specific polyester/cotton gloves with thin nitrile coating on palm and fingertips are flexible and robust offering exceptional grip, dexterity, tactility and durability.
  • Nitrile coating protects the hand from abrasion, slips and penetration of oils, hydrocarbons and grease
  • Absorbs perspiration to increase comfort
  • Antibacterial and anti-odour treatment for greater hygiene
  • Seamless knit to prevent irritation
  • Breathable back of hand to reduce perspiration
  • Designed for easy movement and continuous wear thanks to renowned ergonomic shaped hand mould
Ideal for use in agriculture, construction, logistics, manufacturing, metallurgy, municipal, services and transport. Typical applications include: Handling oily components, heavy duty handling, maintenance, parks and gardens, refuse collection and warehousing.
Certifications: EN 388:2016 (4132X); Cat. II