Imprimer
Glass stoppers, standard ground joint, octagonal, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
216240307 216240401 216241106 216241209 216241303 216241603 216270404 216240804 216240907 216270901 216271306 216240701
217-9122EA 15.1 EUR
217-9122 217-9123 217-9132 217-9134 217-0771 217-0772 217-9143 217-0769 217-0770 217-0775 217-0777 217-0836
Glass stoppers, standard ground joint, octagonal, DURAN®
Bouchons Bouchons en verre
These stoppers are made from borosilicate glass 3.3 and have an octagonal head for good grip.
  • Solid or semi-hollow
Certifications: DIN 12252