Imprimer
Certified thermometers
Fournisseur: Alla France
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
566A100 568J100 568K050 5680J100QPEU4 5680J100QPEU5 5680J100QPEU6 5660A100QPEU4 5660A100QPEU5 5660A100QPEU6
ALLA566A100EA 199 EUR
ALLA566A100 ALLA568J100 ALLA568K050 ALLA5680J100QPEU4 ALLA5680J100QPEU5 ALLA5680J100QPEU6 ALLA5660A100QPEU4 ALLA5660A100QPEU5 ALLA5660A100QPEU6
Certified thermometers
Thermomètres Thermomètres analogiques
These certified thermometers are manufactured using a type N 16 soda lime glass with diameter ~6 to 7 mm.
  • Graduation lines are fine and easily readable
  • The colour of the glass is standardised in yellow giving a better reading than the white back
  • MRA traceable thermometers
The graduation is printed and fused in the glass with a special process and so the graduation resists to all chemical products except fluoridric acid and hot soda (to a lesser extent) because these two attack the glass. They are calibrated for total immersion.
They possess a suspension ring and an expansion chamber to support temperatures higher than those of the thermometer.
Informations de commande: Can be supplied with 'EU' Traceable Calibration Certificate or 'EO' Official Calibration Certificate.
Informations de livraison: Delivered with conformity and traceability certificate.