Imprimer
Glass Stoppers with Octagonal Grip
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0.300.014.BR 0.300.019.BR 0.300.034.BR 0.300.045.BR 0.300.060.BR 0300024BR
217-9145EA 7.5 EUR
217-9145 217-9146 217-9152 217-9154 WITG0.300.060.BR WITG0300024BR
Glass Stoppers with Octagonal Grip
Bouchons Bouchons en verre
Solid standard taper stoppers with octagonal grip, made of Borosilicate glass 3.3.
  • Acc. to DIN 12252
* Semi-hollow