Imprimer
Microtome, Manual LS2065
Fournisseur: Dakewe Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DAKE120.012001-00EA 0 EUR
DAKE120.012001-00
Microtome, Manual LS2065
Microtomes et groupes de microtomes Microtomes
Ergonomical one-step injection molded housing (including fixing bottom studs) results in appropriate structure, attractive appearance and convenient operation, and enables to avoid resonance and lower the frictional resistance.
  • Magnetic waste tray enables easy removal for cleaning
  • Designed black to blend into one harmonious whole together with the main machine
  • Precise, comfort, and safe slicing experience
  • Fast trimming function enables fast switch between sectioning and trimming mode
  • Two safety locking mechanism
Guide rails and retainer in mechanical transmission system was made in German, which are so highly precise that enables to slide with even no gap, thus results in precision and longer equipment life and free of lubrication, maintenance and noise.