Imprimer
Stirrer guides
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.460.029 1.470.029 1.468.004 1.468.029 1.475.029
WITG1.460.029EA 93.1 EUR
WITG1.460.029 WITG1.470.029 WITG1.468.004 WITG1.468.029 WITG1.475.029
Stirrer guides
Agitateurs Outils d'agitation
Stirrer guides are of different types.
With fused-on stirrer bearing.
GTS-vacuum-stirrer, with PTFE-insert and interchangeable PTFE-vacuum-seal, screw cap GL 25.
With PP-insert, 8 mm Ø and screw cap GL 25, made of melamine.
With cooling jacket and PTFE-insert, with interchangeable PTFE-vacuum seal and screw cap GL 25, made of melamine.