Antibody labelling kits, Mix-n-Stain™ Digoxigenin
Marques : BiotiumTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Mix-n-Stain™ Digoxigenin antibody labelling kits utilise a technology that allows rapid antibody labelling with digoxigenin without a purification step.
- Tolerates many common buffer components and antibody stabilisers