Imprimer
Antibody labelling kits, Mix-n-Stain™ DNP
Marques : Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
92326 92327 92325
92326.EA 224 EUR
92326. 92327. 92325.
Antibody labelling kits, Mix-n-Stain™ DNP
Systèmes de liaison et de marquage des protéines
Mix-n-Stain™ DNP antibody labelling kits utilise a technology that allows rapid antibody labelling with dinitrophenol without a purification step.
  • The labelling procedure tolerates many common buffer components and antibody stabilisers