Imprimer
ESD safety ankle boots, lace-up, Wellmaxx, Timothy, GTX
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
769752/36 769752/37 769752/38 769752/39 769752/40 769752/41 769752/42 769752/43 769752/44 769752/45 769752/46 769752/47 769752/48
ELTN769752/36EA 116.9 EUR
ELTN769752/36 ELTN769752/37 ELTN769752/38 ELTN769752/39 ELTN769752/40 ELTN769752/41 ELTN769752/42 ELTN769752/43 ELTN769752/44 ELTN769752/45 ELTN769752/46 ELTN769752/47 ELTN769752/48
ESD safety ankle boots, lace-up, Wellmaxx, Timothy, GTX
Chaussures et bottes
Metal and leather-free ESD safety ankle boots made from hydrophobised microfibre and hydrophobised textile material, Gore-Tex® lining, a full padded bellows tongue and a dual-density PU/PU sole WELLMAXX TRAINERS.
  • Gore-Tex® climate membrane
  • Cold insulating and waterproof
  • Full-length inlay sole ESD PRO black
  • ESD soft-fleece insole with integrated omissions
  • Anatomically shaped insole and ankle support for improved stability
  • Cushioning midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving the energy back to the wearer
  • Protective composite toecap (200 J)
Ideal for use in cold environments and in areas where exposure to moisture is expected.
Colour: Black/yellow
Certifications: EN 61340-5-1 (ESD); EN ISO 20345; S2, SRC, CI