Imprimer
Sheets for ÄKTA Systems, Benchkote®, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2300-10060 2300-10061 2300-10062 2300-10064 2300-10065 2300-10063 2300-10072 2300-10073 2300-10074 2300-10092 2300-10093 2300-10094
WHAT2300-10060EA 19 EUR
WHAT2300-10060 WHAT2300-10061 WHAT2300-10062 WHAT2300-10064 WHAT2300-10065 WHAT2300-10063 WHAT2300-10072 WHAT2300-10073 WHAT2300-10074 WHAT2300-10092 WHAT2300-10093 WHAT2300-10094
Sheets for ÄKTA Systems, Benchkote®, Whatman™
Surface Protectors Bench Protectors
These Benchkote® sheets for ÄKTA systems will protect the top buffer trays from buffer spillages and salt deposits.
  • Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems
Benchkote® material has a high quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.