Imprimer
Disposable knives, SteriWare®
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8039H-100S 8039A-100 8039H-100
SAMP8039H-100SEA 127 EUR
SAMP8039H-100S SAMP8039A-100 SAMP8039H-100
Disposable knives, SteriWare®
Couteaux
These single use knives are made from food grade plastic (100% virgin HDPE). Ideal for sample preparation and general laboratory use.
  • Can be used to cut or split products
  • Available non sterile or gamma irradiated sterile (SAL 10-6)
  • Moulded and packed in a cleanroom
  • No cleaning is required before or after sampling
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • BSE/TSE-free
Certifications: Material conforms to FDA and EU 10/2011. Full batch traceability.