Imprimer
Acrodisc® One PSF Syringe Filters, wwPTFE Membrane
Fournisseur: Cytiva (Formerly Pall Lab)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AP-4910 AP-4912 AP-4913 AP-4915 AP-4916 AP-4918 AP-4919 AP-4921 AP-4917 AP-4914 AP-4920 AP-4916TC
514-1084EA 213 EUR
514-1084 514-1092 514-1086 514-1093 514-1088 514-1094 514-1090 514-1095 514-1219 514-1180 514-1181 514-1456
Acrodisc® One PSF Syringe Filters, wwPTFE Membrane
Filtres Filtres-seringue
The new Acrodisc® One and Acrodisc® PSF premium syringe filters combine the benefits of a multi-layered prefiltration device with an automation-certified filter housing.
  • Increased surface area for maximising throughput
  • Heat sealed membrane for reduced breakthrough
  • Accurate analysis. HPLC certified for low levels of UV-absorbing extractables
  • Available in 0,2 µm for UHPLC applications and with GxF (Glass Fiber) pre-filter for difficult to filter samples
  • The Acrodisc® One offers a Universal Membrane for all your analytical filtration requirements
PP housing with female Luer inlet and male slip outlet connections.
Acrodisc ONE with water wettable PTFE (wwPTFE) has a Next Generation Universal Membrane for all sample types with fast flow rates, low protein binding, and low levels of UV-absorbing extractables. Avoid the expense and inconvenience of keeping a variety of filters on hand, it is a versatile filter for aqueous and aggressive organic solvent-based solutions.
The Acrodisc® One and Acrodisc® PSF GxF syringe filters provide two to four times the throughput of standard prefilter devices for particulate-laden samples.
For ion chromatography specific syringe filters, look for the Acrodisc® ion chromatography syringe filters page.
Certifications: Acrodisc® One PSF syringe filters are Automation Certified to ensure smooth operation and worry-free performance 24 hours a day in automated workstations. HPLC certified for low levels of UV-absorbing extractables.
Cdt: Syringe filters packed in cases of 200 are available in four packs of 50 or in an AutoPack™ format. The AutoPack™ format consists of eight tubes with 25 syringe filters stacked on top of each other.