Imprimer
Aniline Blue diammonium salt, certified_Biological Stain Commission Stain_histology, Sigma-Aldrich®
Marques : SIGMA ALDRICH MICROSCOPY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MQ200
415049-25G 415049-50G
415049-25GEA 112 EUR
415049-25G 415049-50G
Aniline Blue diammonium salt, certified_Biological Stain Commission Stain_histology, Sigma-Aldrich®
Aniline Blue diammonium salt
Designed for use in Mallory′s connective tissue stain and Gomori′s one-step trichrome.
  • Dye content - 65%
  • Absorption, λmax - 605 nm
Formule: C₃₇H₃₂N₅O₉S₃
Poids moléculaire: 786.87 g/mol
Numéro CAS: 66687-07-8