Imprimer
Total immersion thermometers, solid stem
Fournisseur: Amarell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
G10500-FL G10506-FL G10510-FL
AMARG10500-FLEA 29.9 EUR
AMARG10500-FL AMARG10506-FL AMARG10510-FL
Total immersion thermometers, solid stem
Thermomètres Thermomètres analogiques
These glass thermometers are suitable for standard everyday use.
  • White backing
  • Reliable measurements and solid glass
Ø: 5,5 to 7,0 mm.