Imprimer
Seal test, Stericlin®
Fournisseur: VP Medical Packaging
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3FSZB830106 3FSZB830108
489-0147EA 185 EUR
489-0147 489-0148
Seal test, Stericlin®
Leak Detection Systems Liquid Leak Detection Systems
The Stericlin® Seal Test is used for routine testing of seal seams according to DIN EN ISO 11607. In combination with the seal seam integrity test it offers the most comprehensive and reliable routine examination for the heat sealing process.
  • Clear result
  • Show irregularities and damage
For specifications, see the DGSV guidelines for the validation of packaging processes according to DIN EN ISO 11607-2.