Imprimer
Electrodes for surfactant titration
Fournisseur: METROHM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6.0507.120 6.0507.010
METR6.0507.120EA 1890 EUR
METR6.0507.120 METR6.0507.010
Electrodes for surfactant titration
Électrodes Électrodes, autres
Surfactant electrodes for two-phase titration.
  • Long working life with normal use
  • Installation length of 125 mm
  • Minimum immersion depth is 20 mm
  • Metrohm plug-in head G
Ionic surfactant electrode for the titration of anionic and cationic surfactants in aqueous matrices. Make response due to ionophores immobilised in the membrane.
NIO surfactant electrode for the titration of non-ionic surfactants in aqueous matrices, for the titration of surfactants based on polyoxyethylene adducts, for the titration of pharmaceutical ingredients.