Imprimer
Ductless filtering fume hoods, with external connection, LABOPUR®
Fournisseur: TRIONYX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BH06D BH06Z BH09D BH09Z BH12D BH12Z BH15D BH15G BH15Z
TRIOBH06DEA 1870 EUR
TRIOBH06D TRIOBH06Z TRIOBH09D TRIOBH09Z TRIOBH12D TRIOBH12Z TRIOBH15D TRIOBH15G TRIOBH15Z
Ductless filtering fume hoods, with external connection, LABOPUR®
Fume Hoods and Enclosures Ductless Fume Hoods
These fume hoods are intended to be connected to an exhausting system for the discharge of toxic and harmful vapours to the outside of the building.
  • Steel construction with white epoxy paint RAL 9010 which is highly resistant to chemicals
  • Ergonomic hand traps for safe and easy handling in the enclosure
  • Hands passage or trapezoidal opening
  • Ideal for storage of flammables, corrosives and toxics
Delivered ready to use, no assembly required and to be connected only to the electrical network and to an exhausting system. Transparent glass walls, made of PMMA, on all sides offer optimal illumination on the work top and immediate visualisation of the products handled. Lids allow the introduction of cables for power supply of equipment and fluids inside the ductless fume hood.