Imprimer
PCR tube strips with attached caps, EasyStrip™ Plus
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AB-2000 AB-2005
ADVAAB-2000EA 391 EUR
ADVAAB-2000 ADVAAB-2005
PCR tube strips with attached caps, EasyStrip™ Plus
Tubes Tubes PCR
Thermo Scientific™ EasyStrip™ Plus tube strip with attached caps is designed to reduce the risk of cross-contamination in PCR and qPCR reactions. The tube strip is compatible with 0,2 ml thermal cycler blocks and features individually attached optical caps for quick and easy access to individual samples. In addition, graduated 20 µl markers on each tube enable visual volume verification and measurement of reagent and sample additions.
  • Avoid cross contamination with the individually attached caps
  • Verify tube reaction volumes with graduated 20 μl markings on each tube
  • Track samples using the end tabs to label each strip
  • Identify individual wells quickly and prevent pipetting errors using the labels (A-H) on each tube cap and cap hinge (visible when cap is open and closed)
  • Available with flat caps for PCR and optically clear caps for qPCR