Imprimer
PCR tube strips, Eppendorf FAST
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0030124.901 0030124.910 0030124.928
EPPE0030124.901EA 180 EUR
EPPE0030124.901 EPPE0030124.910 EPPE0030124.928
PCR tube strips, Eppendorf FAST
Tubes Tubes PCR
Eppendorf Fast PCR tube strips made of polyethylene transfer heat faster than tubes made of traditional plastics. The Fast PCR tube strips are particularly suited to scientists using Fast PCR reagent kits, scientists trying to speed up protocols or cyclers with high heating and cooling rates.
  • Ideal for use with Fast cyclers and Fast enzymes
  • 8 reaction tubes in strip-format – ideal for small sample volumes
  • Easily adaptable for automation
  • Sealable using flat or domed strip lids
  • Certified free from human DNA, DNase, RNase and PCR inhibitors
The Fast PCR Tube Strip allows scientists using Fast cyclers (such as the Eppendorf Mastercycler X50) and Fast enzymes to obtain higher PCR yield. This is achieved by the faster heat transfer from block to reaction, allowing for better control over the reaction. The tube strips are suitable for Fast end-point PCR and Real-Time PCR applications.