Imprimer
DNA transfection reagent, broad spectrum, Helix-IN™
Marques : OZ BIOSCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HX10100 HX10500 HX11000
OZBIHX10100EA 78.48 EUR
OZBIHX10100 OZBIHX10500 OZBIHX11000
DNA transfection reagent, broad spectrum, Helix-IN™
Réactifs de transfection
Helix-IN™ DNA transfection reagent opens up new possibilities for addressing issues of classical transfection technologies. Helix-IN™ is a patented Cationic Hydroxylated Amphiphilic Multi-block Polymer (CHAMP™ Technology). This novel bi-functional co-polymer is biocompatible, ionisable, pH responsive
  • Broad spectrum transfection reagent for cell lines and hard-to-transfect cells
  • High transfection efficiency and increased transgene expression even with low DNA amount
  • High intracellular protein production while preserving viability
  • High secreted protein production while minimising cellular stress
  • Avoid secondary effects
  • Compatible with any culture medium
Informations de livraison: Each kit contains one vial of Helix-IN reagent plus one vial of HIB Enhancer reagent.