Imprimer
Seal seam integrity test, Stericlin®
Fournisseur: VP Medical Packaging
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3FSZB830102 3FSZB830103 3FSZB830104
489-0144EA 158 EUR
489-0144 489-0145 489-0146
Seal seam integrity test, Stericlin®
Détecteurs de fuites Détecteurs de fuites de liquides
Stericlin® seal seam integrity tests are used for routine testing of seal seams according to DIN EN ISO 11607. In combination with the Seal Test they offer comprehensive and secure routine testing for the heat sealing process.
  • Easy to perform
  • Quickly detects channels and voids
For specifications, see the DGSV guidelines for the validation of packaging processes according to DIN EN ISO 11607-2.
Informations de commande: Seal seam integrity tests are available in two basic versions: 'Pipette' and 'disposable ink bag'.