Imprimer
Shoe covers, non slip, PP
Fournisseur: Hubei Orient International
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
114-9102EA 104 EUR
114-9102 114-9100 114-9099 114-9103
Shoe covers, non slip, PP
Vêtements à usage unique
Single-use over shoes made from PP feature a latex coated anti-slip sole.
  • Soft and comfortable
  • Skid-free soles for extra traction and improved safety
Suitable for food processing, manufacturing, construction, hygienic application and daily use.
Weight: 40 g/m²
Cdt: 10 pieces/roll, 10 rolls/polybag, 10 polybags/carton.