Imprimer
Mouse IgM purification kit
Marques : LIGATRAP TECHNOLOGIES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
LT-145 LT-145KIT LT-145-1X1ML LT-145-1X5ML LT-145-3X1ML
LIGALT-145EA 0 EUR
LIGALT-145 LIGALT-145KIT LIGALT-145-1X1ML LIGALT-145-1X5ML LIGALT-145-3X1ML
Mouse IgM purification kit
Protéines purification Protein Purification Kits
LigaTrap® Mouse IgM Purification Kit with LT technology utilises novel, patented ligands, designed for IgM antibody purification. Kit provides all the necessary reagents for fast, convenient micro-scale purification of mouse IgM in just ten easy steps.
  • Designed to purify mouse IgM
  • For in vitro laboratory use
  • Binding capacity for mouse IgM is >10 mg/ml resin
  • Storage: 2 to 8 °C
LigaTrap® mouse IgM purification resin is engineered to purify high quality mouse IgM antibodies from recombinant and monoclonal sources. Resins bind an exceptionally broad range of recombinant, monoclonal and human/animal source polyclonal immunoglobulins.
Each spin column can be used, regenerated, and used up to ten times with minimal
loss in binding capacity.
Informations de livraison: Each kit contains ten microspin columns prefilled with 0,1 ml of LigaTrap® mouse IgM purification resin, buffers and collection tubes.
Attention: Not for diagnostic or therapeutic use. Not for human or animal consumption.