Imprimer
NMR sample tubes, susceptibility matched, Shigemi®, SP Wilmad-LabGlass
Fournisseur: SP Industries
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BMS-003 BMS-005B
WILMBMS-003EA 424 EUR
WILMBMS-003 WILMBMS-005B
NMR sample tubes, susceptibility matched, Shigemi®, SP Wilmad-LabGlass
Tubes Tubes RMN
Susceptibility matched tubes can reduce the sample volume down to 1/3 by minimising the susceptibility gradients occurring at the solvent-air interface.