Imprimer
Heat sealer, Impulse
Fournisseur: TUTTNAUER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TSM260M TSM470M
TUTTTSM260MEA 828 EUR
TUTTTSM260M TUTTTSM470M
Heat sealer, Impulse
Thermosoudeuses Soude sachet
Impulse sealing unit for sealing of medical products. Sterilisation of medical instruments is a critically important activity in medicine and dentistry, as well as in veterinary surgeries, hospitals, clinics and laboratories.
  • Fast and reliable impulse sealers
  • Complies with the strictest standards
Sterilisation is effective only if the sterility of the instruments is preserved from the point of sterilisation until they are used. The sealing process ensures that the instruments remain sealed after sterilisation and maintain perfect sterility during their shelf-life.
Certifications: Compliance with DIN standards 58953-7 and EN 868-4/5.