Imprimer
Precision laboratory thermometers with safety-coating, ACCU-SAFE
Fournisseur: Ludwig Schneider GmbH & CO. KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1042344S/80 1042343S/80 1042346S/80 1042347S/80 1042349S/80 1042351S/80 1042352S/80 1042353S/80 1042354S/80 1042604S/80 1042703S/80 1042704S/80 1122904S/80 1122905S/80 1122906S/80 1122907S/80 1042345S/80
SCNL1042344S/80EA 0 EUR
SCNL1042344S/80 SCNL1042343S/80 SCNL1042346S/80 SCNL1042347S/80 SCNL1042349S/80 SCNL1042351S/80 SCNL1042352S/80 SCNL1042353S/80 SCNL1042354S/80 SCNL1042604S/80 SCNL1042703S/80 SCNL1042704S/80 SCNL1122904S/80 SCNL1122905S/80 SCNL1122906S/80 SCNL1122907S/80 SCNL1042345S/80
Precision laboratory thermometers with safety-coating, ACCU-SAFE
Thermomètres Thermomètres analogiques
These total immersion thermometers are suitable for calibration.
  • Colourless prismatic capillary form
  • Enclosed scale with blue filling
  • High transparency for easy legibility
  • Usable as stirring thermometer
The rugged encapsulation protects against splintered glass or display-fluid leakage. The protective coating is remarkable for its high transparency and quick response time allowing easily legibility. It provides temperature resistance (up to 250 °C) and hence sterilisability and a high degree of acid and alkali resistance (hydrofluoric acid).
Informations de commande: For calibration options, contact your Avantor representative.
Informations de livraison: With individual serial number and certificate of conformity.
Cdt: Each item packed in square plastic sleeve.