Imprimer
ASTM® precision thermometers, Gallium filled
Fournisseur: Amarell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A300030-GA A300040-GA A300050-GA A300060-GA A300110-GA A300120-GA A300130-GA A300140-GA A300170-GA A300180-GA A300190-GA A300200-GA A300460-GA A300620-GA A300808-GA A300810-GA A300960-GA A300970-GA A300980-GA A300990-GA A301000-GA A301010-GA A301100-GA A301110-GA A301120-GA A301230-GA A301240-GA A301250-GA A301260-GA A301380-GA A301390-GA A301410-GA A301420-GA A301430-GA A301440-GA A301450-GA A301470-GA A301490-GA A301940-GA
AMARA300030-GAEA 172 EUR
AMARA300030-GA AMARA300040-GA AMARA300050-GA AMARA300060-GA AMARA300110-GA AMARA300120-GA AMARA300130-GA AMARA300140-GA AMARA300170-GA AMARA300180-GA AMARA300190-GA AMARA300200-GA AMARA300460-GA AMARA300620-GA AMARA300808-GA AMARA300810-GA AMARA300960-GA AMARA300970-GA AMARA300980-GA AMARA300990-GA AMARA301000-GA AMARA301010-GA AMARA301100-GA AMARA301110-GA AMARA301120-GA AMARA301230-GA AMARA301240-GA AMARA301250-GA AMARA301260-GA AMARA301380-GA AMARA301390-GA AMARA301410-GA AMARA301420-GA AMARA301430-GA AMARA301440-GA AMARA301450-GA AMARA301470-GA AMARA301490-GA AMARA301940-GA
ASTM® precision thermometers, Gallium filled
Thermomètres Thermomètres analogiques
These models are suitable for government testing or official calibration.
  • Solid stem thermometer
  • White backed
  • Durable pigment
Certifications: Compliant with ASTM.