Imprimer
Capillary columns, OPTIMA® 5 HT
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
726104.15 726102.15 726102.30 726104.30 726106.15 726106.30 726108.15 726108.30
MANA726104.15EA 628 EUR
MANA726104.15 MANA726102.15 MANA726102.30 MANA726104.30 MANA726106.15 MANA726106.30 MANA726108.15 MANA726108.30
Capillary columns, OPTIMA® 5 HT
Colonnes pour chromatographie
Nonpolar silarylene GC phase with low bleeding and high temperature stability.
  • Ideal for MS detectors
  • Recommended for simulated distillation, hydrocarbons, fuel, oil analysis, and high boiling analytes
  • High temperatures stability with the maximum temperature for isothermal operation of 380 °C
  • Maximum temperature for short isotherms in a temperature program 400 °C
The polarity index is according to 5% diphenyl - 95% dimethylpolysiloxane phases.
USP code: G27/G36.