Imprimer
Capillary columns, OPTIMA® XLB
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
725850.30 725850.60
MANA725850.30EA 1160 EUR
MANA725850.30 MANA725850.60
Capillary columns, OPTIMA® XLB
Colonnes pour chromatographie
Nonpolar silarylene GC phase with low bleeding for envrionmental analysis.
  • Ideal for ion trap and quadrupole MS detectors
  • Highly selective for environmental and trace analyses such as pesticides and PCBs
  • Maximum temperature for isothermal operation 340 °C
  • Maximum temperature for short isotherms in a temperature program 360 °C
Phase with optimized silarylene content.