Imprimer
Reading lamps, pure visionLED
Fournisseur: ESCHENBACH OPTIK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
16030 16034
ESCH16030EA 190 EUR
ESCH16030 ESCH16034
Reading lamps, pure visionLED
Éclairages Éclairage LED
These models are easy-to-operate thanks to a handy tactile switch.
  • Large and comfortable distance to reading material
  • Sturdy arm with flexible and freely movable lampshade
  • Shadow-free and homogeneous illumination – No maintenance or lamp replacement necessary
Colour: Silver/anthracite