Imprimer
Pipette and filter tips for Rainin® LTS® pipettes, xTIP4™, low retention
Fournisseur: BIOTIX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
63300001 63300002 63300003 63300004 63300005 63300006 63300007 63300008 63300009 63300020 63300021 63300022 63300023 63300024 63300015 63300016 63300018 63300019 63300931
732-3424EA 163 EUR
732-3424 732-3425 732-3426 732-3427 613-5951 613-5952 613-5953 613-5954 613-5955 613-5956 613-5957 613-5958 613-5959 613-5960 613-5961 613-5962 613-5964 613-5965 732-3483
Pipette and filter tips for Rainin® LTS® pipettes, xTIP4™, low retention
Pointes de pipettes
xTIP4™ is the only Rainin® LTS® compatible tip engineered with StarStop, a patented positive stop feature. This feature minimizes ejection force and reduces the risk of repetitive stress injuries (RSI).
  • FlexFit sealing technology lowers the forces necessary to attach and eject tips with a secure seal
  • All tips are Low binding to maximize sample recovery and increase accuracy
  • Blade tip reduces the need for tip touch off and increases accuracy and precision
  • Compatible with both multi- and single channel Rainin® LTS® pipettes
  • Filter tips are made from polyethylene with a 99,99% bacterial filter efficiency (BFE)
FlexFit®, X-Resin®, and Blade® technologies developed by Biotix are found exclusively in Biotix and xTIP brand pipette tips. Biotix technologies minimise inaccuracies associated with the number one issue in pipetting: user error. These proven features are standard on all xTIP low retention pipette tips, maximising accuracy and precision.
Tip Eject® reloads: An easy to use manual ejection reload that loads tips without user contact to the snapcard giving a cleaner refill transfer. Protected in a transparent sleeve for easy tip identification and protection from environmental particulates. Compact design uses 1/5 of the bench space and 80% less plastic when compared to racked product. TipEject™ is 100% recyclable.
Certifications: Manufactured under stringent controls in an ISO 9001:2015 certified facility. Certified free of DNase/RNase, endotoxins, pyrogens, ATP, and PCR inhibitors.
Informations de livraison: Supplied with color coded snapcards by volume and printed with part number, lot number and description for high traceability.