Imprimer
HPLC columns, PAH analysis, NUCLEODUR® C18 PAH
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
760784.40 760786.40 760773.30 761970.30 760786.30 761971.30 760773.20 760773.40 760783.30 760783.40 760784.30 760785.30 760785.40 761970.20
MANA760784.40EA 1170 EUR
MANA760784.40 MANA760786.40 MANA760773.30 MANA761970.30 MANA760786.30 MANA761971.30 MANA760773.20 MANA760773.40 MANA760783.30 MANA760783.40 MANA760784.30 MANA760785.30 MANA760785.40 MANA761970.20
HPLC columns, PAH analysis, NUCLEODUR® C18 PAH
Colonnes pour chromatographie
Special octadecyl modified silica phase for HPLC analysis of PAHs.
  • High purity spherical silica gel
  • Polymeric octadecyl modification
  • Allows efficient gradient separation of PAHs according to EPA and EFSA
NUCLEODUR® C18 PAH columns are recommended for a gradient separation of 16 PAHs in accordance with EPA. An ultra-fast separation of 16 EPA PAHs is possible with 1,8 µm UHPLC phase. Moreover, separations of further PAH compounds such as PAHs according to EFSA can also be realised.
Selectivity: matched for PAH analysis.
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).