Imprimer
Malt diastase, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0210153925 0210153980
ICNA0210153925EA 56.9 EUR
ICNA0210153925 ICNA0210153980
Malt diastase, MP Biomedicals
Enzymes
DIASTASE from malt, 1X N.F. stable, 60 mesh powder. Ready for CE IVD labeling of clinical applications.
  • White to tan powder
  • Soluble in water
  • Store at 2 to 8 °C
Attention: For research use only (RUO).