Imprimer
Rescue system, R350, Rollgliss™ DBI-SALA®, 3M™
Fournisseur: CAPITAL SAFETY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AG6350ST31/60 AG6350ST51/60
CPSDAG6350ST31/60EA 0 EUR
CPSDAG6350ST31/60 CPSDAG6350ST51/60
Rescue system, R350, Rollgliss™ DBI-SALA®, 3M™
Protections anti-chutes Kits de prévention des chutes
The Rollgliss R350 system provides efficient hauling, but more importantly, very controlled lowering (a finger and thumb are all that is required to hold a load). This system also allows to change the hauling ratios on the fly with quick release pulleys, allowing custom rigging for different applications and jobsites.
  • Interchangeable pulley system with a 3:1 ratio standard
  • Rope retention bar does not allow rope to twist or overlap
  • Visual and audible function controls
  • Static kernmantle rope for optimum performance
  • Weight capacity of 150 kg
Quick connect system allows fast insertion of various pulleys.
Certifications: EN 341, EN 1496