Imprimer
Extendible pole hoist system, DBI-SALA, 3M™
Fournisseur: CAPITAL SAFETY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8560409 8560476
CPSD8560409EA 0 EUR
CPSD8560409 CPSD8560476
Extendible pole hoist system, DBI-SALA, 3M™
Protections anti-chutes Cordes de sécurité
This pole hoist attaches to a carabiner on the universal bracket to a suitable anchor point, allowing to work in any direction from any angle.
  • Swivel head that rotates 360 degrees, providing unparalleled flexibility in both vertical and horizontal confined-space entries
  • Overall length adjusts for positioning attendant for working in close quarters
Certifications: EN 795 Class B
Informations de livraison: Includes extendible pole hoist, swivel head and stainless steel hardware, also a winch bracket.